LIF007178901 Life Chemicals Inc.

N-(3-Chloro-phenyl)-phthalamic acid

  • MDL number MFCD00029949
  • Cas Number : 5406-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart