LIF007178904 Life Chemicals Inc.

4-[(3-Chlorophenyl)amino]-4-oxobutanoic acid

  • MDL number MFCD00449641
  • Cas Number : 15386-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart