LIF007178918 Life Chemicals Inc.

4-[(5-Methyl-4-phenyl-1,3-thiazol-2-yl)amino]-4-oxobutanoic acid

  • MDL number MFCD01438849
  • Cas Number : 325471-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart