LIF007179331 Life Chemicals Inc.

(2-Imino-1,3-thiazolidin-3-yl)acetic acid hydrobromide

  • MDL number MFCD02250829
  • Cas Number : 479192-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart