LIF007179342 Life Chemicals Inc.

2-Amino-6-methyl-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carboxylic acid amide

  • MDL number MFCD00617279
  • Cas Number : 95211-68-0
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart