LIF007179631 Life Chemicals Inc.

2-(3-methylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

  • MDL number MFCD00187753
  • Cas Number : 67822-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart