LIF007179742 Life Chemicals Inc.

(4-Methyl-quinolin-2-yl)-hydrazine

  • MDL number MFCD00818533
  • Cas Number : 21703-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart