LIF007179776 Life Chemicals Inc.

4-Acetyl-3,4-dihydro-1H-quinoxalin-2-one

  • MDL number MFCD01995115
  • Cas Number : 120589-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart