LIF007179870 Life Chemicals Inc.

[(4-Methylphenyl)sulfonyl]acetonitrile

  • MDL number MFCD00039487
  • Cas Number : 5697-44-9
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart