LIF007179960 Life Chemicals Inc.

1-Amino-3-(4-tert-butyl-phenoxy)-propan-2-ol

  • MDL number MFCD00447828
  • Cas Number : 41403-84-3
  • Molecular Weight : 223.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart