LIF007179962 Life Chemicals Inc.

1-Amino-3-(2,3-dihydro-1,4-benzodioxin-2-ylmethoxy)propan-2-ol hydrochloride

  • MDL number MFCD04426765
  • Cas Number : 1049784-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart