LIF007180021 Life Chemicals Inc.

1-Phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-ol

  • MDL number MFCD00082637
  • Cas Number : 21314-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart