LIF007180042 Life Chemicals Inc.

2-(4-Methylphenyl)-2,6-dihydro-4H-thieno[3,4-c]pyrazol-3-amine

  • MDL number MFCD03030150
  • Cas Number : 858668-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart