LIF007180044 Life Chemicals Inc.

2-(4-Chloro-phenyl)-2,6-dihydro-4H-thieno[3,4-c]pyrazol-3-ylamine

  • MDL number MFCD03030145
  • Cas Number : 214542-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart