LIF007180055 Life Chemicals Inc.

5-(3,4-Dimethoxy-phenyl)-[1,3,4]thiadiazol-2-ylamine

  • MDL number MFCD00981427
  • Cas Number : 5427-87-2
  • Molecular Weight : 237.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart