LIF007180087 Life Chemicals Inc.

1-(4-Chloro-phenyl)-1,2,3,4-tetrahydro-pyrrolo[1,2-a]pyrazine

  • MDL number MFCD01605570
  • Cas Number : 112758-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart