LIF007180114 Life Chemicals Inc.

2-(4-Fluorophenyl)-2,6-dihydro-4H-thieno[3,4-c]pyrazol-3-amine

  • MDL number MFCD04440708
  • Cas Number : 865546-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart