LIF007180118 Life Chemicals Inc.

6-amino-5-nitropyrimidin-4-ol

  • MDL number MFCD01912544
  • Cas Number : 36746-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart