LIF007180197 Life Chemicals Inc.

3-(4-Chlorophenyl)-2-mercapto-6,7-dihydrothieno[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD04440754
  • Cas Number : 887201-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart