LIF007180213 Life Chemicals Inc.

(2-{[2-(4-Methoxyphenyl)-1H-indol-3-yl]thio}ethyl)amine

  • MDL number MFCD04440769
  • Cas Number : 865546-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart