LIF007180373 Life Chemicals Inc.

3-((4-chlorophenyl)thio)propanoic acid

  • MDL number MFCD02636303
  • Cas Number : 6310-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart