LIF007223991 Life Chemicals Inc.

2-Amino-5-nitro-thiophene-3-carboxylic acid ethyl ester

  • MDL number MFCD01566377
  • Cas Number : 42783-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart