LIF007244570 Life Chemicals Inc.

4-(Chloromethyl)-5,7-dimethyl-2H-chromen-2-one

  • MDL number MFCD03664464
  • Cas Number : 41295-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart