LIF007251537 Life Chemicals Inc.

(Quinolin-8-ylthio)acetic acid

  • MDL number MFCD02659656
  • Cas Number : 122179-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart