LIF007266165 Life Chemicals Inc.

4-(tert-butyl)-6-chloropyrimidine

  • MDL number MFCD03731166
  • Cas Number : 3435-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart