LIF007266219 Life Chemicals Inc.

6-Fluoro-2-methylquinoline-3-carboxylic acid

  • MDL number MFCD03085774
  • Cas Number : 461026-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart