LIF007266236 Life Chemicals Inc.

3-Bromo-4-nitropyridine

  • MDL number MFCD03085767
  • Cas Number : 89364-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart