LIF007266238 Life Chemicals Inc.

2-Bromo-4,6-dimethyl-pyridin-3-ylamine

  • MDL number MFCD03731181
  • Cas Number : 104829-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart