LIF007266244 Life Chemicals Inc.

4-Hydroxy-nicotinic acid

  • MDL number MFCD00040286
  • Cas Number : 609-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart