LIF007276562 Life Chemicals Inc.

2-Chloro-6-methyl-4-phenyl-quinazoline

  • MDL number MFCD00436573
  • Cas Number : 5185-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart