LIF007277891 Life Chemicals Inc.

2-Amino-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carbonitrile

  • MDL number MFCD00128278
  • Cas Number : 4651-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart