LIF007277951 Life Chemicals Inc.

2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

  • MDL number MFCD00121495
  • Cas Number : 70291-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart