LIF007278055 Life Chemicals Inc.

2,6-Dichloro-4-phenylquinoline

  • MDL number MFCD00436567
  • Cas Number : 10352-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart