LIF007279193 Life Chemicals Inc.

1-Adamantan-1-yl-ethanol

  • Cas Number : 26750-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart