LIF007279239 Life Chemicals Inc.

3-Amino-adamantane-1-carboxylic acid hydrochloride

  • MDL number MFCD01825827
  • Molecular Weight : 231.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart