LIF007279573 Life Chemicals Inc.

3-Acetyl-4-phenyl-1H-quinolin-2-one

  • MDL number MFCD00226511
  • Cas Number : 313273-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart