LIF007279890 Life Chemicals Inc.

5-amino-2-methoxybenzonitrile

  • MDL number MFCD00723841
  • Cas Number : 214623-57-1
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart