LIF007280500 Life Chemicals Inc.

Diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate

  • MDL number MFCD00005450
  • Cas Number : 4815-30-9
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart