LIF007281423 Life Chemicals Inc.

1,3-Dihydroxy-naphthalene-2-carboxylic acid ethyl ester

  • MDL number MFCD00451682
  • Cas Number : 6843-89-6
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart