LIF007285317 Life Chemicals Inc.

2-Amino-thiophene-3-carboxylic acid amide

  • MDL number MFCD00706455
  • Cas Number : 14080-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart