LIF007285675 Life Chemicals Inc.

2-Chloro-4-phenyl-quinazoline

  • MDL number MFCD01152724
  • Cas Number : 29874-83-7
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart