LIF007294697 Life Chemicals Inc.

(5-Nitro-1H-benzimidazol-2-yl)methanol

  • MDL number MFCD00462582
  • Cas Number : 20034-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart