LIF007338630 Life Chemicals Inc.

Furan-2-yl-(1H-imidazol-2-yl)-methanone

  • MDL number MFCD02669592
  • Cas Number : 62457-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart