LIF007338668 Life Chemicals Inc.

(6-Bromo-2-methoxy-1-naphthyl)acetic acid

  • MDL number MFCD02669590
  • Cas Number : 329269-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart