LIF007344343 Life Chemicals Inc.

1-(1H-Benzoimidazol-2-yl)-ethanol

  • MDL number MFCD00016718
  • Cas Number : 19018-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart