LIF007347038 Life Chemicals Inc.

(5,7-Dimethyl-[1,2,4]triazolo[4,3-a]pyrimidin-3-ylsulfanyl)-acetic acid

  • MDL number MFCD01960597
  • Cas Number : 41266-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart