LIF007373250 Life Chemicals Inc.

4-(1H-Benzoimidazol-2-yl)-1-phenyl-pyrrolidin-2-one

  • MDL number MFCD03085143
  • Cas Number : 380561-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart