LIF007374150 Life Chemicals Inc.

2-(2-Phenoxy-acetylamino)-3-phenyl-propionic acid

  • MDL number MFCD00266828
  • Cas Number : 110728-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart