LIF007377008 Life Chemicals Inc.

1-(2,4-Dihydroxyphenyl)-2-(2-methoxyphenyl)ethanone

  • MDL number MFCD00205538
  • Cas Number : 92549-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart