LIF009172614 Life Chemicals Inc.

1,2,3,3a-Tetrahydropyrrolo[1,2-a]quinoxalin-4(5H)-one

  • MDL number MFCD01098256
  • Cas Number : 21550-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart